M9251
3-Methyl-3-ethylglutarimide
powder
Synonym(s):
3-Ethyl-3-methylglutarimide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
EC Number:
MDL number:
UNSPSC Code:
12352100
form
powder
color
white
mp
126-127 °C (lit.)
Storage temp.
−20°C
SMILES string
CCC1(C)CC(=O)NC(=O)C1
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) antagonists: from bench to bedside.
Henri Mattes et al.
Journal of medicinal chemistry, 53(15), 5367-5382 (2010-04-02)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service