Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8391

Sigma-Aldrich

Methyl Purple

Synonym(s):

Methyl Purple indicator solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C67H62N8Na4O14S3
CAS Number:
Molecular Weight:
1391.41
MDL number:
UNSPSC Code:
12171500

SMILES string

CN(C)c1ccc(cc1)N=Nc2ccccc2C(=O)O[Na].CN(C)c3ccc(cc3)N=Nc4ccccc4C(=O)O[Na].CCN(Cc5cccc(c5)S(=O)(=O)O[Na])c6ccc(cc6)C(=C7/C=CC(C=C7)=[N+](CC)Cc8cccc(c8)S([O-])(=O)=O)\c9ccc(O)cc9S(=O)(=O)O[Na]

InChI

1S/C37H36N2O10S3.2C15H15N3O2.4Na/c1-3-38(24-26-7-5-9-33(21-26)50(41,42)43)30-15-11-28(12-16-30)37(35-20-19-32(40)23-36(35)52(47,48)49)29-13-17-31(18-14-29)39(4-2)25-27-8-6-10-34(22-27)51(44,45)46;2*1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20;;;;/h5-23H,3-4,24-25H2,1-2H3,(H3,41,42,43,44,45,46,47,48,49);2*3-10H,1-2H3,(H,19,20);;;;/q;;;4*+1/p-4/b;2*17-16+;;;;

InChI key

CNJVTYXLOYTWPD-HFZRWZJVSA-J

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service