Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6671

Sigma-Aldrich

Methyl 4,7,8,9-tetra-O-acetyl-2-thio-N-acetylneuraminic acid methyl ester

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103

assay

≥98%

Storage temp.

−20°C

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hasegawa, A., et al.
Journal of Carbohydrate Chemistry, 10, 493-493 (1991)
A facile, large-scale preparation of the methyl 2-thioglycoside of N-acetylneuraminic acid, and its usefulness for the alpha-stereoselective synthesis of sialoglycosides.
A Hasegawa et al.
Carbohydrate research, 212, 277-281 (1991-06-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service