Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L6252

Sigma-Aldrich

Lys-Val hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23N3O3 · HCl
CAS Number:
Molecular Weight:
281.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

Cl.CC(C)C(NC(=O)C(N)CCCCN)C(O)=O

InChI

1S/C11H23N3O3.ClH/c1-7(2)9(11(16)17)14-10(15)8(13)5-3-4-6-12;/h7-9H,3-6,12-13H2,1-2H3,(H,14,15)(H,16,17);1H

InChI key

OKIYIEZRMCQJCX-UHFFFAOYSA-N

Application

Lysylvaline (Lys-Val) may be used to study supramolecular packing crystal structures of dipeptides.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Recacha et al.
The journal of peptide research : official journal of the American Peptide Society, 50(5), 388-392 (1997-12-24)
The title compound crystallizes in the zwitterionic form. The crystal forms a supramolecular structure with the peptide molecules organized in head-to-tail columns in the b direction. The arginine side-chains and acetate ions interact with neighbor peptides in the c direction.
J Soman et al.
International journal of peptide and protein research, 32(5), 352-360 (1988-11-01)
L-Lysine D-glutamate crystallizes in the monoclinic space group P2(1) with a = 4.902, b = 30.719, c = 9.679 A, beta = 90 degrees and Z = 4. The crystals of L-lysine D-aspartate monohydrate belong to the orthorhombic space group
H P Yennawar et al.
International journal of peptide and protein research, 38(6), 569-573 (1991-12-01)
L-Valyl-L-lysine hydrochloride, C11N3O3H23 HCl, crystallizes in the monoclinic space group P2(1) with a = 5.438(5), b = 14.188(5), c = 9.521(5) A, beta = 95.38(2) degrees and Z = 2. The crystal structure, solved by direct methods, refined to R
G A Belokrylov et al.
International journal of immunopharmacology, 21(12), 879-883 (1999-12-22)
In experiments on mice and the influence in vitro of the dipeptides GluTrp, LysAsp, LysVal, AspGlu, GluAsp on their constituent amino acids and mixtures on the indexes of specific and non-specific resistance was studied. There is no difference between the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service