Skip to Content
MilliporeSigma
All Photos(1)

Documents

L3010

Sigma-Aldrich

1-Hexanoyl-sn-glycero-3-phosphocholine

~99%

Synonym(s):

L-α-Lysophosphatidylcholine, caproyl, Lysolecithin, caproyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H30NO7P
CAS Number:
Molecular Weight:
355.36
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

~99%

storage temp.

−20°C

SMILES string

CCCCCC(=O)OC[C@H](O)COP(O)(=O)OCC[N](C)(C)C

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service