Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

KEM0019

Sigma-Aldrich

T4 DNA Ligase (Rapid)

Ultra-pure enzyme for nucleic acid modifications

Sign Into View Organizational & Contract Pricing


About This Item

grade

for molecular biology

assay

>99% (SDS-PAGE)

form

buffered aqueous solution

specific activity

300,000 U/mg

concentration

600,000 U/mL

shipped in

dry ice

storage temp.

−20°C

General description

T4 DNA Ligase catalyzes the formation of a phosphodiester bond between the terminal 5′ phosphate and a 3′ hydroxyl groups of duplex DNA or RNA. The enzyme efficiently joins blunt and cohesive ends and repairs single stranded nicks in duplex DNA, RNA or DNA/RNA hybrids.

Application

Suitable for:
  • Restriction cloning
  • TA cloning
  • Adapter ligation
  • NGS library construction
  • Other applications requiring high efficiency ligation

Features and Benefits

  • Ultra-purification process for ultimate enzyme performance
  • Highest quality specifications for ultimate product consistency
  • Undetectable DNA and nuclease contamination

Components

Supplied with:KEM0046B (2X Rapid Ligation Buffer)KEM0049B (10X T4 DNA Ligase Buffer)

Unit Definition

1 unit is defined as the amount of DNA Ligase required to join 50% of 100 ng of DNA fragments with cohesive termini in 50 μL 1X DNA Ligase Buffer following a 30 minute incubation at 23° C.

Physical form

Supplied in 10 mM Tris-HCl, 50 mM KCl, 1 mM DTT, 0.1 mM EDTA and 50% glycerol at pH 7.5 @ 25° C.

Other Notes

Source of protein: A recombinant E. coli strain carrying the T4 DNA Ligase gene.
Unit size: 240,000 U

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laure Rittié et al.
Journal of cell communication and signaling, 2(1-2), 25-45 (2008-09-04)
Since molecular cloning has become routine laboratory technique, manufacturers offer countless sources of enzymes to generate and manipulate nucleic acids. Thus, selecting the appropriate enzyme for a specific task may seem difficult to the novice. This review aims at providing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service