Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I6765

Sigma-Aldrich

D-myo-Inositol 1,5,6-tris­phosphate ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15O15P3 · 6H3N
CAS Number:
Molecular Weight:
522.28
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

storage temp.

−20°C

SMILES string

N.O[C@H]1[C@@H](O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@@H]1O

InChI

1S/C6H15O15P3.H3N/c7-1-2(8)4(19-22(10,11)12)6(21-24(16,17)18)5(3(1)9)20-23(13,14)15;/h1-9H,(H2,10,11,12)(H2,13,14,15)(H2,16,17,18);1H3/t1-,2-,3-,4+,5-,6-;/m1./s1

InChI key

MWPGDKRESDTMSH-TYZYTZLSSA-N

Other Notes

Found in avian erythrocytes.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L R Stephens et al.
The Biochemical journal, 265(2), 435-452 (1990-01-15)
Avian erythrocytes were incubated with myo-[3H]inositol for 6-7 h and with [32P]Pi for the final 50-90 min of this period. An acid extract was prepared from the prelabelled erythrocytes, and the specific radioactivities of the gamma-phosphate of ATP and of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service