Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I3753

Sigma-Aldrich

8-Hydroxy-7-iodo-5-quinolinesulfonic acid

Synonym(s):

Ferron, Iodoxyquinolinesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6INO4S
CAS Number:
Molecular Weight:
351.12
Beilstein/REAXYS Number:
223832
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

269-270 °C (dec.) (lit.)

SMILES string

Oc1c(I)cc(c2cccnc12)S(O)(=O)=O

InChI

1S/C9H6INO4S/c10-6-4-7(16(13,14)15)5-2-1-3-11-8(5)9(6)12/h1-4,12H,(H,13,14,15)

InChI key

ZBJWWKFMHOAPNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service