Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA008726

Sigma-Aldrich

Anti-ZNF132 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc finger protein 132 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EHQGTQSEEKPYTCGACGRDFWLNANLHQHQKEHSGGKPFRWYKDRDALMKSSKVHLSENPFTCREGGKVILGSCDLLQLQAVDSGQKPYSNLGQLPEVCTTQKLFECSNCGKAFLKSSTLPNHLRTHSE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF132(7691)

Related Categories

General description

ZNF132 (zinc finger protein 132) is a member of the Cys2-His2 (C2H2) zinc finger protein family. This gene is localized to human chromosome 19q13.4. This protein is predicted to contain one Kruppel-associated box domain and 18 C2H2 zinc finger domains.

Immunogen

Zinc finger protein 132 recombinant protein epitope signature tag (PrEST)

application

Anti-ZNF132 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

The functional characteristics of ZNF132 (zinc finger protein 132) are not yet defined, but its structure suggests that it might play a role in the suppression of transcription. The gene locus that it is localized to is generally deleted in thyroid adenomas. The expression of this gene is significantly suppressed in prostate cancer (PC), and this is linked to high Gleason score and advanced T stage. The down-regulation of this gene in PC is an outcome of its hypermethylation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70916

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mette Opstrup Abildgaard et al.
International journal of cancer, 130(4), 885-895 (2011-03-30)
This study investigates the expression and biomarker potential of zinc finger protein 132 (ZNF132) in prostate cancer (PC) by transcriptional profiling and immunohistochemical analysis of tissue microarrays, including tumor specimens from 615 radical prostatectomy (RP) patients and 199 conservatively treated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service