Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA001808

Sigma-Aldrich

Anti-SAV1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-WW45, Anti-WWP4, Anti-salvador, Anti-salvador homolog 1 (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$508.30

$508.30

List Price$598.00Save 15%

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$508.30

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$508.30

List Price$598.00Save 15%

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

IGRVAATSLGNLTNHGSEDLPLPPGWSVDWTMRGRKYYIDHNTNTTHWSHPLEREGLPPGWERVESSEFGTYYVDHTNKKAQYRHPCAPSVPRYDQPPPVTYQPQQTERNQSLLVPANPYHTAEIPDWLQVYARAPVKYDHILKWELFQLA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SAV1(60485)

Immunogen

salvador family WW domain containing protein 1

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SAV1, a component of the Hippo pathway, is a molecule associated with the regulation of both cell proliferation and cell death.[1] It is composed of two WW domains and a coiled-coil region. It has been found to be expressed in almost all human cell lines and localizes in both the cytoplasm and nucleus. Previously it was reported that, SAV1 conjugates with the mammalian sterile 20-like kinase 1 (MST1) and forms heterodimer, which plays a major role in MST1-driven cell death signaling pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70365

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Ewa Kowalczyk et al.
Cancers, 15(24) (2023-12-23)
Salvador homolog-1 (SAV1) is a component of the Hippo pathway that regulates tissue growth and homeostasis by affecting diverse cell processes, including apoptosis, cell division, and differentiation. The aberrant expression of Hippo pathway components has been observed in various human
Keiko Matsuura et al.
BMC cancer, 11, 523-523 (2011-12-22)
Clinical outcome of patients with high-grade ccRCC (clear cell renal cell carcinoma) remains still poor despite recent advances in treatment strategies. Molecular mechanism of pathogenesis in developing high-grade ccRCC must be clarified. In the present study, we found that SAV1
Xuelai Luo et al.
International journal of molecular medicine, 23(3), 357-362 (2009-02-13)
Mammalian sterile 20-like kinase 1 (MST1) is a serine/threonine protein kinase that is activated in response to a variety of apoptotic stimuli and causes apoptosis when over-expressed in mammalian cells. The physiological regulation and cellular targets of MST1 are not
Nam Jin Yoo et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 111(6), 595-598 (2003-09-13)
It is believed that clonal expansion and cancer growth is the result of the deregulation of proliferation and cell death. Recently, salvador, a molecule that can regulate both cell proliferation and cell death, was identified. It was also reported that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service