Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H8910

Sigma-Aldrich

Heat Shock Cognate 70-interacting Protein from rat

≥70% (SDS-PAGE), recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

Quality Level

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

buffered aqueous solution

concentration

1.1 mg protein/mL

UniProt accession no.

shipped in

dry ice

Storage temp.

−70°C

Gene Information

Biochem/physiol Actions

Hsc70 (heat shock cognate 70) is a molecular chaperone and constitutively expressed type of Hsp70 (heat shock protein 70). It participates in synaptic signal transduction. In hippocampal neurons, it can interact with NFκB (nuclear factor κB) p65. In presence of oxidative stress, it shows protective effect on cardiomyocytes, partly via association with α-enolase. Hsp70 and Hsc70 together are involved in the production and degradation of ERAD (endoplasmic reticulum-associated degradation) substrates. Hsc70 is also needed for myofibril structure maintenance and the suppression of myofibrillar degeneration in presence of mechanical stress.

Physical form

Solution in 20 mM MOPS, pH 7.2, 50 mM KCl, and 2 mM MgCl2

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihiro Matsumura et al.
Molecular biology of the cell, 22(16), 2797-2809 (2011-06-24)
The Hsp/c70 cytosolic chaperone system facilitates competing pathways of protein folding and degradation. Here we use a reconstituted cell-free system to investigate the mechanism and extent to which Hsc70 contributes to these co- and posttranslational decisions for the membrane protein
Akinori Hishiya et al.
Circulation research, 107(10), 1220-1231 (2010-10-05)
A homozygous disruption or genetic mutation of the bag3 gene, a member of the Bcl-2-associated athanogene (BAG) family proteins, causes cardiomyopathy and myofibrillar myopathy that is characterized by myofibril and Z-disc disruption. However, the detailed disease mechanism is not yet
Christin Klenke et al.
PloS one, 8(6), e65280-e65280 (2013-06-14)
Signaling via NF-κB in neurons depends on complex formation with interactors such as dynein/dynactin motor complex and can be triggered by synaptic activation. However, so far a detailed interaction map for the neuronal NF-κB is missing. In this study we
Qi Luo et al.
Free radical research, 45(11-12), 1355-1365 (2011-10-01)
Constitutive heat shock protein 70 (Hsc70) is a molecular chaperone that has been shown to protect cardiomyocytes against oxidative stress. However, the molecular mechanism responsible for this protection remains uncertain. To understand the mechanism associated with the myocardial protective role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service