Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H4534

Sigma-Aldrich

Entero-hylambatin

≥95% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H91N17O18S2
Molecular Weight:
1438.63
MDL number:
UNSPSC Code:
12352200

assay

≥95% (HPLC)

form

powder

color

white

solubility

H2O: soluble

storage temp.

−20°C

Amino Acid Sequence

Asp-Pro-Pro-Asn-Pro-Asp-Arg-Phe-Tyr-Gly-Met-Met-NH2

General description

Entero-hylambatin is a tachykinin peptide. Tachykinins are mainly neuropeptides.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative Physiology and Evolution of the Autonomic Nervous System
Nilsson C B
CRC Press, 4, 81-85 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service