Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1329-01

Bromophenol Blue

Cytiva 17-1329-01, pack of 10 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.47

form

solid

feature

wetted part: no

packaging

pack of 10 g

manufacturer/tradename

Cytiva 17-1329-01

storage condition

room temperature

color

Purple, black

pH range

3.0-4.6, yellow to purple

solubility

cold water: soluble (Soluble in cold water and hot water)

General description

PlusOne Bromophenol Blue is an electrophoresis tracking dye used in IEF, PAGE and Sequencing.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes components of rehydration solutions for use in 2-D Electrophoresis with Cytiva products.

Protocols

This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service