Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G5643

Sigma-Aldrich

6-O-β-D-Galactopyranosyl-D-galactose

≥95%

Synonym(s):

β-D-Gal-(1→6)-D-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

assay

≥95%

storage temp.

−20°C

SMILES string

OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O

Application

Useful for binding studies of immunoglobulins specific for β-D-(1→6)-linked D-galactopyranans.[1][2]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The binding of 5-(dimethylamino)-1-naphthalenesulfonyl (dansyl) derivatives of d-galactose saccharides to several immunoglobulins capable of binding beta-(1 leads to 6)-linked D-galactopyranans.
A K Bhattacharjee et al.
Carbohydrate research, 67(2), 536-540 (1978-12-01)
Glaudemans, C.P.J., et al.
Carbohydrate Research, 40, 129-129 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service