Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F5926

Supelco

Fillalbin

≥97% (TLC), powder, white

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO4
Molecular Weight:
291.34
MDL number:
UNSPSC Code:
41116107

assay

≥97% (TLC)

form

powder

color

white

storage temp.

2-8°C

Biochem/physiol Actions

Causes cramps; increases arterial blood pressure and breathing frequency.

Warning

Protect from light.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service