Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F1133

Sigma-Aldrich

Fmoc-Lys(Boc)-OH

Synonym(s):

Nα-Fmoc-Nε-Boc-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N2O6
CAS Number:
Molecular Weight:
468.54
Beilstein/REAXYS Number:
4217767
EC Number:
MDL number:
UNSPSC Code:
12352200

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C26H32N2O6/c1-26(2,3)34-24(31)27-15-9-8-14-22(23(29)30)28-25(32)33-16-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h4-7,10-13,21-22H,8-9,14-16H2,1-3H3,(H,27,31)(H,28,32)(H,29,30)/t22-/m0/s1

InChI key

UMRUUWFGLGNQLI-QFIPXVFZSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald Domalaon et al.
Amino acids, 46(11), 2517-2530 (2014-07-30)
Taking a minimalistic approach in efforts to lower the cost for the development of new synthetic antimicrobial peptides, ultrashort cationic lipopeptides were designed to mimic the amphiphilic nature crucial for their activity but with only a very short peptide sequence
Yun Song et al.
Cell reports, 30(8), 2699-2711 (2020-02-27)
The transcriptional corepressor complex CoREST is one of seven histone deacetylase complexes that regulate the genome through controlling chromatin acetylation. The CoREST complex is unique in containing both histone demethylase and deacetylase enzymes, LSD1 and HDAC1, held together by the
Yunfei Sun et al.
International journal of pharmaceutics, 468(1-2), 83-90 (2014-04-09)
Gene therapy is a potent tool for the treatment of cancer and other gene defect diseases, which involves using DNA that encodes a functional, therapeutic gene to replace a mutated gene. However, the DNA transfection efficiency is restricted by its
Soo Min Cho et al.
Scientific reports, 4, 6777-6777 (2014-10-28)
Amyloid-β (Aβ) is one of the few neuropathological biomarkers associated with transporters of the blood-brain barrier (BBB). Despite the well-characterized clinical indication of decreasing Aβ levels in the cerebrospinal fluid (CSF) during the development of Alzheimer's disease (AD), the link
Paul W R Harris et al.
Biopolymers, 104(2), 116-127 (2015-02-07)
The cancer protein NY-ESO-1 has been shown to be one of the most promising vaccine candidates although little is known about its cellular function. Using a chemical protein strategy, the 180 amino acid polypeptide, tagged with an arginine solubilizing tail

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service