EPAG2-10
Platform for Eppendorf® New Brunswick™ Innova® 2000 Platform Shaker
for 10 mL Erlenmeyer flasks, dedicated, aluminum
Select a Size
$763.00
Select a Size
About This Item
$763.00
Recommended Products
material
aluminum
feature
dedicated
for 10 mL Erlenmeyer flasks
manufacturer/tradename
Eppendorf® AG2-10 (Global)
L × W
76 cm × 46 cm
capacity
(60 x 10 mL Erlenmeyer flasks)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service