Documents
EP951022109
Eppendorf® Masterclear cap strips and real-time PCR tube strips
pack of 120 ea (10 x 12 strips of 8 tubes & caps)
About This Item
UNSPSC Code:
41121800
material
polypropylene
sterility
non-sterile
packaging
pack of 120 ea (10 x 12 strips of 8 tubes & caps)
manufacturer/tradename
Eppendorf® 951022109
capacity
0.1 mL
color
clear wells
white border
General description
Masterclear® Cap Strips and real-time PCR Tube Strips, 120 pcs. (10 × 12 pcs.)
  • real-time PCR Tube Strips
  • White wells for better reflection, high mechanical stability
  • Extremely thin walls for optimal heat transfer
  • Certified free from human DNA, DNase, RNase and PCR inhibitors1)
  • 8 reaction tubes in strip-format for 0.1 mL – ideal for small sample volumes
  • Masterclear Cap Strips
  • Inverted dome to avoid scratching of the optic surface, inverted dome to reduce volume of the tubes
  • Optimized for maximum light transmission
  • Certified free from human DNA, DNase, RNase and PCR inhibitors1)
  • Strips with eight microcaps for 0.1 and 0.2 mL wells
1) Download of certificates under www.eppendorf.com/certificates
Features and Benefits
  • Inverted dome to avoid scratching of the optic surface
  • Inverted dome to reduce volume of the tubes
  • Optimized for maximum light transmission
  • Certified free from human DNA, DNase, RNase and PCR inhibitors1)
  • Strips with eight microcaps for 0.1 and 0.2 mL wells
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Masterclear is a registered trademark of Eppendorf AG