Key Documents
EP5490041003
Rotor for Eppendorf® Concentrator Plus
F-50-8-16, non aerosol tight, to hold, 8 x 8.0/12.0 mL round-bottom tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Product EP5490041003 is not currently sold in your country. Contact Technical Service
feature
to hold 8 x 16 x 105 -120 mm
to hold 8 x 8.0/12.0 mL round-bottom tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490041003 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Concentrator Plus
General description
Selection of rotors for the Eppendorf Concentrator Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)