E6782
Anti-ERGIC-53/p58−Cy3 antibody produced in rabbit
1.5-3.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-ERGIC53, Anti-F5F8D, Anti-FMFD1, Anti-LMAN1, Anti-MCFD1, Anti-MR60, Anti-gp58
About This Item
Recommended Products
biological source
rabbit
conjugate
CY3 conjugate
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human, mouse
concentration
1.5-3.0 mg/mL
technique(s)
direct immunofluorescence: 0.5-1.0 μg/mL using human HeLa cells
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... LMAN1(3998)
mouse ... Lman1(70361)
rat ... Lman1(116666)
Application
Western Blotting (1 paper)
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
related product
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service