Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E6517

Sigma-Aldrich

8,11-Eicosadiynoic Acid

≥98%, ethanol solution

Synonym(s):

EDYA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O2
CAS Number:
Molecular Weight:
304.47
MDL number:
UNSPSC Code:
12352204

assay

≥98%

form

ethanol solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCCC#CCC#CCCCCCCC(O)=O

Biochem/physiol Actions

Non-toxic acetylenic fatty acid that inhibits eicosanoid biosynthesis at several stages. Inhibits arachidonyl synthase, acyl-CoA synthase, cyclooxygenase, and arachidonate uptake by platelets.

Warning

Oxygen and light sensitive.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service