E1137
Ecgonine methyl ester-(N-methyl-d3) hydrochloride
Synonym(s):
(−)-3β-Hydroxy-1αH-5αH-tropane-2β-carboxylic acid methyl ester, 3-Hydroxy-8-(methyl-::FId::FN3)-8-azabicyclo[3.2.1]octane-2-carboxylic acid methyl ester
About This Item
isotopic purity
98 atom % D
drug control
USDEA Schedule II; Home Office Schedule 2; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
food and beverages
format
neat
InChI
1S/C10H17NO3.ClH/c1-11-6-3-4-7(11)9(8(12)5-6)10(13)14-2;/h6-9,12H,3-5H2,1-2H3;1H/t6-,7?,8?,9+;/m0./s1
InChI key
RLLMNWXOZDXKCQ-JVVAZNKBSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service