Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D4400

SAFC

Diethyl chlorophosphate

Synonym(s):

Diethyl phosphorochloridate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)Cl
CAS Number:
Molecular Weight:
172.55
Beilstein/REAXYS Number:
471433
EC Number:
MDL number:
UNSPSC Code:
12352200

vapor density

5.94 (vs air)

vapor pressure

0.1 mmHg ( 25 °C)

refractive index

n20/D 1.416 (lit.)

bp

60 °C/2 mmHg (lit.)

density

1.194 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOP(Cl)(=O)OCC

InChI

1S/C4H10ClO3P/c1-3-7-9(5,6)8-4-2/h3-4H2,1-2H3

InChI key

LGTLXDJOAJDFLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Acute Tox. 3 Inhalation

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

199.4 °F - closed cup

flash_point_c

93 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service