Skip to Content
MilliporeSigma

D3566

Anti-DcR1 antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Decoy Receptor 1, Anti-LIT, Anti-TRAIL-R3, Anti-TRID

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~65 kDa

species reactivity

mouse, rat, human

concentration

~0.5 mg/mL

technique(s)

microarray: suitable, western blot: 0.5-1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TNFRSF10C(8794)

General description

DcR1 is found in most normal human tissue, but not in most cancer cell lines. It is a member of the TNF family of TRAIL (TNF-related apoptosis-induced ligand) receptors.

Immunogen

synthetic peptide corresponding to amino acids 111-123 at the extracellular domain of human DcR1 precursor.

Application

Anti-DcR1 antibody produced in rabbit is suitable for immunoblotting at a working concentration of 0.5-1μg/mL using human HeLa cells and mouse and rat liver tissue lysates.

Biochem/physiol Actions

DcR1 is a glycosylphosphatidylinositol-linked membrane protein that binds TRAIL (APO2 ligand). It acts as a decoy receptor and binds TRAIL to its extracellular TRAIL-binding domain, thus inhibiting TRAIL-mediated apoptosis. It does not have an intracellular death domain and, therefore, does not induce apoptosis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library