Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D104

Sigma-Aldrich

4-Diphenylacetoxy-N-methylpiperidine methiodide

powder

Synonym(s):

4-DAMP methiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26INO2
CAS Number:
Molecular Weight:
451.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

color

white

solubility

H2O: 3 mg/mL

SMILES string

[I-].C[N+]1(C)CCC(CC1)OC(=O)C(c2ccccc2)c3ccccc3

Gene Information

Biochem/physiol Actions

Muscarinic acetylcholine receptor antagonist with selectivity for the M3 amd M5 subtypes.

Caution

Photosensitive

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Moriya et al.
Life sciences, 64(25), 2351-2358 (1999-06-22)
A family of five subtypes of muscarinic acetylcholine receptors (mAChR) has been identified based on their molecular structures and second signal transduction pathways. In the present study, we examined the antagonist binding profiles of 9 muscarinic antagonists (atropine, 4-DAMP, pirenzepine
N Watson et al.
British journal of pharmacology, 127(2), 590-596 (1999-06-29)
1. Affinity estimates were obtained for several muscarinic antagonists against carbachol-stimulated [3H]-inositol phosphates accumulation in Chinese hamster ovary (CHO-KI) cells stably expressing either human muscarinic M3 or M5 receptor subtypes. The rationale for these studies was to generate a functional
Jing Hu et al.
Journal of ethnopharmacology, 139(1), 136-141 (2011-11-24)
Crotonis Fructus is the mature fruit of Croton tiglium L. (Euphorbiaceae), which has been used in traditional Chinese medicine for the treatment of gastrointestinal (GI) diseases, such as constipation, abdominal pain, peptic ulcer, and intestinal inflammation for thousands of years.
T M Cembala et al.
British journal of pharmacology, 125(5), 1088-1094 (1998-12-10)
1. Neuromuscular blocking drugs (NMBD's) are known to produce cardiovascular side effects manifesting as brady/tachycardias. In this study we have examined the interaction of a range of steroidal NMBD's with recombinant human m1-m5 muscarinic receptors expressed in Chinese hamster ovary
Yiming Zhang et al.
Cellular signalling, 23(6), 1057-1067 (2011-02-19)
Cobrotoxin (CbT), a short-chain postsynaptic α-neurotoxin, has been reported to play a role in analgesia. However, to date, the detailed mechanisms still remain unknown. In the present study, we identify a novel functional role of CbT in modulating T-type Ca(2+)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service