Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0145

Sigma-Aldrich

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane

≥90%

Synonym(s):

TIPDSiCl2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2Si(Cl)OSi(Cl)[CH(CH3)2]2
CAS Number:
Molecular Weight:
315.43
Beilstein/REAXYS Number:
1934216
MDL number:
UNSPSC Code:
12352000

assay

≥90%

refractive index

n20/D 1.454 (lit.)

bp

70 °C/0.5 mmHg (lit.)

density

0.986 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)[Si](Cl)(O[Si](Cl)(C(C)C)C(C)C)C(C)C

InChI

1S/C12H28Cl2OSi2/c1-9(2)16(13,10(3)4)15-17(14,11(5)6)12(7)8/h9-12H,1-8H3

InChI key

DDYAZDRFUVZBMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

168.8 °F

flash_point_c

76 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service