CN350-05
Canine Coronary Artery Smooth Muscle Cells: CnCASMC (Cryovial)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
canine coronary artery (normal, tunica intima and media)
packaging
pkg of 500,000 cells
manufacturer/tradename
Cell Applications, Inc
karyotype
2n = 78
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cardiovascular diseases
shipped in
dry ice
storage temp.
−196°C
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Canine Coronary Artery Smooth Muscle Cells (CnCASMC) provide an excellent animal model system to study all aspects of cardiovascular function and disease.
Canine Coronary Artery Smooth Muscle Cells (CnCASMC) provide an excellent animal model system to study all aspects of cardiovascular function and disease.
Cell Line Origin
Artery
Subculture Routine
Please refer to the CnCASMC Culture Protocol.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service