Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

YTS 154.7.7.10

Reactivity: Anti-Thy-1, mouse, 87072285

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Rat x Rat Hybridoma

ImmunoDon

DA rat spleen

Myeloma

Y3Ag1.2.3

Reactivity

Anti-Thy-1, mouse

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Monoclonal antibody mouse CD90 (Thy-1)

Cell Line Description

The antibody is directed against the monomorphic determinant of the mouse Thy-1 T cell antigen. Has been used in T+NK cell depletion in vivo and flow cytometry studies. Cambridge Enterprise Reference number AB10:www.enterprise.cam.ac.uk.

Immunogen

CBA mouse thymocytes

Culture Medium

RPMI 1640 + 2mM glutamine + 0.05mM 2-mercaptoethanol (2ME) + 20% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9 x 105 cells/ml. 5% CO2; 37°C. Saturation density 9 x 105/ml

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service