Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C2523

Sigma-Aldrich

Cholestane-3β,5α,6β-triol

Synonym(s):

3β,5α,6β-Trihydroxycholestane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H48O3
CAS Number:
Molecular Weight:
420.67
MDL number:
UNSPSC Code:
12352200

SMILES string

CC(C)CCC[C@@H](C)C1CCC2C3CC(O)[C@@]4(O)CC(O)CCC4(C)C3CCC12C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alzbeta Kloudova-Spalenkova et al.
The Journal of steroid biochemistry and molecular biology, 197, 105566-105566 (2019-12-25)
Oxygenated metabolites of cholesterol (oxysterols) have been previously demonstrated to contribute to progression of various cancers and to modulate resistance to breast cancer endocrine therapy in vitro. We measured prognostic roles of circulating levels of seven major oxysterols in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service