Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-12576

Sigma-Aldrich

Human Uterine Smooth Muscle Cells (HUtSMC)

500,000 proliferating cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human uterus (myometrium)

packaging

pkg of 500,000 cells

morphology

(smooth muscle)

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

room temp

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Uterus

Application

Primary Human Uterine Smooth Muscle Cells (HUtSMC) are isolated from the myometrium (the middle layer of the uterine wall) and are provided in a proliferating format. HUtSMC stain positive for smooth muscle α-actin. During pregnancy, hormones and growth factors cause an extensive hypertrophy of uterine smooth muscle cells. This and other morphological and biochemical effects on the uterus can be studied in vitro using HUtSMC.

Quality

Rigid quality control tests are performed for each lot of Human Smooth Muscle Cells. They are tested for cell morphology, adherence rate, and cell viability. Flow cytometric analyses for cell-type specific markers, e.g. smooth muscle a-actin, are carried out for each lot. Growth performance is tested through multiple passages up to 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 7500 - 10000 cells per cm2Passage After Thawing: P2Tested Markers: Smooth muscle alpha-actin positiveGuaranteed population doublings: >15

Recommended products

Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Smooth muscle human primary cells isolated from blood vessels and hollow organs, plus smooth muscle cell growth media. Protocols on media handling, cell passaging and product use.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service