Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B4375

Sigma-Aldrich

Nα-Benzoyl-L-argininamide hydrochloride monohydrate

Synonym(s):

BAA hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19N5O2 · HCl · H2O
CAS Number:
Molecular Weight:
331.80
EC Number:
UNSPSC Code:
12352200

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Miwa et al.
Biochemical and biophysical research communications, 112(2), 754-762 (1983-04-29)
The 55- (H-UK) and 36-kDa forms (L-UK) of human urinary urokinase lost most of esterase activity toward acetyl-glycyl-L-lysine methyl ester upon reductive cleavage of 3 SS bonds with dithiothreitol in the presence of the competitive inhibitor, N alpha-benzoyl-L-arginine amide (BAA)
R M Epand et al.
Biophysical chemistry, 19(4), 355-362 (1984-06-01)
The effects of an amino acid derivative (N-benzoyl-L-argininamide), four small peptides (Phe-Gly-Phe-Gly, gastrin-related peptide (Trp-Met-Arg-Phe-NH2), tetragastrin (Trp-Met-Asp-Phe-NH2), pentagastrin (Boc-beta Ala-Trp-Met-Asp-Phe-NH2] and one medium-sized peptide, glucagon (29 residues), on the gel-to-liquid crystalline transition of a multilamellar suspension of dimyristoylphosphatidylcholine have been
S Kunugi et al.
Biochimica et biophysica acta, 704(1), 107-113 (1982-05-21)
The effects of pressure on the trypsin-catalyzed hydrolyzed hydrolyses of three specific substrates, N-benzoyl-L-arginine ethyl ester (BzArgOEt), amide (BzArgNH2) and p-nitroanilide (BzArgNA), have been examined. The volume of the activation (delta V++) for kcat was -2.4 ml/mol for BzArgOEt and
N Miwa et al.
Biochimica et biophysica acta, 791(1), 1-8 (1984-11-23)
The disulfide bond in Cys-194-Cys-222 of the isolated B chain (UK X B) of human urinary urokinase was selectively carboxamidomethylated (R X CAM-UK X B) after specific reduction with dithiothreitol in the presence of the competitive inhibitor N-alpha-benzoyl-L-argininamide (BzArgNH2) and
D F Hare et al.
Experimental parasitology, 68(2), 168-175 (1989-02-01)
The proteinase activity of Giardia lamblia trophozoites, Portland 1 strain, was characterized with respect to substrate specificities and inhibitor sensitivities. Proteinase activity with urea-denatured hemoglobin (UDH), alpha-N-benzoyl-DL-arginine-2-naphthylamide (BANA), and alpha-N-benzoyl-argininamide (BAA) as substrates exhibited pH optima of 5.8, 3.8, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service