Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2656

Sigma-Aldrich

Benextramine tetrahydrochloride

≥75% (HPLC), solid

Synonym(s):

BHC, Dithiobis(N-[N-(2-methoxybenzyl)-6-aminohexyl]-2-aminoethane), Dithiobis(N-[N-(2-methoxybenzyl)-6-aminohexyl]-2-aminoethane);, N,N′′-(Dithiodi-2,1-ethanediyl)bis[N′-(2-methoxyphenylmethyl)-1,6-hexanediamine] dihydrochloride, N,N′-Bis(6-[o-Methoxybenzylamino]hexyl)cystamine, N,N′-Bis(6-[o-methoxybenzylamino]hexyl)cystamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H54N4O2S2 · 4HCl
CAS Number:
Molecular Weight:
736.77
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

≥75% (HPLC)

form

solid

color

white

solubility

H2O: soluble (Aqueous solutions are stable for at least one week at −20°C)
ethanol: soluble (warm)
methanol: soluble

storage temp.

2-8°C

SMILES string

Cl.COc1ccccc1CNCCCCCCNCCSSCCNCCCCCCNCc2ccccc2OC

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Adrenoceptor antagonist that differentiates between
two α adrenergic receptor subtypes by binding irreversibly at α1 adrenoceptors, but reversibly to α2 adrenoceptors. Irreversible Neuropeptide Y antagonist.

Physical form

Hygroscopic, photosensitive solid

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Horike et al.
Naunyn-Schmiedeberg's archives of pharmacology, 356(6), 756-762 (1998-02-07)
Several physiological stimuli cause a rise in intracellular Ca2+ concentration ([Ca2+]i) in cardiomyocytes. This increased [Ca2+]i must be restored to physiological resting level to ensure response to further stimuli. In the present study, we examined the effect of neuropeptide Y
P H Van der Graaf et al.
European journal of pharmacology, 300(3), 211-214 (1996-04-11)
We have studied the effects of benextramine on the U46619 (11 alpha,9 alpha-epoxymethano-15S-hydroxy-prosta-5Z,13E-dienoic acid)-mediated contraction of the rat isolated small mesenteric artery. U46619 (10 nM-10 microM) produced a concentration-dependent contraction of the small mesenteric artery. The selective prostanoid TP receptor
J K Phillips et al.
British journal of pharmacology, 124(7), 1403-1412 (1998-09-02)
1. We have investigated the neurotransmitters and receptor subtypes involved in nerve-mediated vasoconstriction in small arteries of the rat hepatic mesentery. 2. A dense sympathetic innervation was demonstrated using catecholamine histochemistry and antibodies against the synaptic vesicle protein synaptophysin. 3.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service