Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV36332

Sigma-Aldrich

Anti-AKAP7 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-A kinase (PRKA) anchor protein 7, Anti-AKAP18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKAP7(9465)

General description

AKAP7 is a member of the A-kinase anchoring protein family. It contains a RII-binding domain allowing it to bind to the regulatory subunit (RII) of cAMP-dependent protein kinase A (PKA) targeting it to the cellular membrane or cytoskeletal structures. It has a molecular weight of 36 kDa.

Immunogen

Synthetic peptide directed towards the middle region of human AKAP7

Biochem/physiol Actions

AKAP7 is a member of the A-kinase anchoring protein (AKAP) family, a group of functionally related proteins that bind to a regulatory subunit (RII) of cAMP-dependent protein kinase A (PKA) and target the enzyme to specific subcellular compartments. AKAPs have a common RII-binding domain, but contain different targeting motifs responsible for directing PKA to distinct intracellular locations. Three alternatively spliced transcript variants encoding different isoforms have been described. Additional variants exist, but their full-length natures have not been determined.This gene encodes a member of the A-kinase anchoring protein (AKAP) family, a group of functionally related proteins that bind to a regulatory subunit (RII) of cAMP-dependent protein kinase A (PKA) and target the enzyme to specific subcellular compartments. AKAPs have a common RII-binding domain, but contain different targeting motifs responsible for directing PKA to distinct intracellular locations. Three alternatively spliced transcript variants encoding different isoforms have been described. Additional variants exist, but their full-length natures have not been determined.

Sequence

Synthetic peptide located within the following region: MKLSKSPWLRKNGVKKIDPDLYEKFISHRFGEEILYRIDLCSMLKKKQSN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abderrahmane Bengrine et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 21(4), 1189-1201 (2007-01-25)
Protein kinase A anchoring proteins or AKAPs regulate the activity of many ion channels. Protein kinase A (PKA) is a well-recognized target of AKAPs, with other kinases now emerging as additional targets. We examined the roles of epithelial-expressed AKAPs in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service