Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7920

Sigma-Aldrich

Aluminum sulfate hexadecahydrate

ACS reagent, 98.0-102.0% (as the labeled hydrate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · 16H2O
CAS Number:
Molecular Weight:
630.40
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

assay

98.0-102.0% (as the labeled hydrate)

composition

water, 14-18 mol/mol

impurities

≤0.01% Insoluble matter
≤0.2% Substances not precipitated by ammonium hydroxide (as SO4)

anion traces

chloride (Cl-): ≤0.005%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.001%

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Al.3H2O4S.16H2O/c;;3*1-5(2,3)4;;;;;;;;;;;;;;;;/h;;3*(H2,1,2,3,4);16*1H2/q2*+3;;;;;;;;;;;;;;;;;;;/p-6

InChI key

TYYRFZAVEXQXSN-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service