Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7877

Sigma-Aldrich

L-Alanine benzyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2 · HCl
CAS Number:
Molecular Weight:
215.68
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

assay

≥98% (TLC)

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

Cl.CC(N)C(=O)OCc1ccccc1

InChI

1S/C10H13NO2.ClH/c1-8(11)10(12)13-7-9-5-3-2-4-6-9;/h2-6,8H,7,11H2,1H3;1H

InChI key

RLMHWGDKMJIEHH-UHFFFAOYSA-N

Application

L-Alanine benzyl ester (ALAB) may be used as an organic reagent and as a cationic charge inducer in microemulsions.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tayyaba Syed et al.
Journal of enzyme inhibition and medicinal chemistry, 26(5), 668-680 (2011-01-22)
2-(1-[(4-Chloro/methylphenylsulfonylamino)alkyl]-5-thioxo-4,5-dihydro-1,3,4-oxadiazoles (4a-e) were synthesized, in four steps, via the sulfonyl derivatives of l-amino acids (l-alanine, l-methionine and l-phenylalanine) 1a-e, the esters 2a-e, the hydrazides 3a-e and finally the cyclization to 4a-e. Alkylation of 4a-e with 1.0 mole eq. of substituted
M S Jie et al.
Lipids, 35(10), 1135-1145 (2000-12-05)
Primary amines (ammonia, methyl, propyl, octyl, octadecyl, phenyl, benzyl, phenethyl) including methyl esters of amino acids (glycine, DL-alanine, L-valine, L-leucine, L-tyrosine, and L-methionine), and secondary amines (dimethyl, diethyl, dipropyl, diisopropyl, dioctyl, and diphenyl) attack regiospecifically the central carbon atom of
Elena Peira et al.
International journal of pharmaceutics, 346(1-2), 119-123 (2007-07-10)
The study reports pig-skin permeation and skin accumulation of miconazole nitrate (MCZ) from positively charged microemulsions containing water, 1-decanol/1-dodecanol (2:1, w/w), lecithin and/or decyl polyglucoside at different weight ratios, propylene glycol, 1,2 hexanediol and a cationic charge-inducing agent (stearylamine (ST)
Jose Manuel Ageitos et al.
Biomacromolecules, 17(1), 314-323 (2015-12-02)
The chemoenzymatic polymerization of amino acid monomers by proteases involves a two-step reaction: the formation of a covalent acyl-intermediate complex between the protease and the carboxyl ester group of the monomer and the subsequent deacylation of the complex by aminolysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service