Skip to Content
MilliporeSigma
All Photos(1)

Documents

A7625

Sigma-Aldrich

N-Acetyl-3,5-dinitro-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11N3O8
CAS Number:
Molecular Weight:
313.22
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

[H][C@@](Cc1cc(c(O)c(c1)[N+]([O-])=O)[N+]([O-])=O)(NC(C)=O)C(O)=O

InChI

1S/C11H11N3O8/c1-5(15)12-7(11(17)18)2-6-3-8(13(19)20)10(16)9(4-6)14(21)22/h3-4,7,16H,2H2,1H3,(H,12,15)(H,17,18)/t7-/m0/s1

InChI key

CFKZKLOBRPCKTF-ZETCQYMHSA-N

pictograms

Flame over circle

signalword

Warning

hcodes

Hazard Classifications

Ox. Sol. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service