Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A5137

Sigma-Aldrich

3-Acetylpyridine adenine dinucleotide, reduced form

90-95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N6O14P2
CAS Number:
Molecular Weight:
662.44
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

assay

90-95%

form

solid

storage temp.

−20°C

SMILES string

[H]P(=O)(OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23)OP([O-])(=O)OC[C@@H]4O[C@@H]([C@@H](O)[C@H]4O)[n+]5cccc(c5)C(C)=O

InChI

1S/C22H28N6O14P2/c1-10(29)11-3-2-4-27(5-11)21-17(32)15(30)12(40-21)6-38-43(34,35)42-44(36,37)39-7-13-16(31)18(33)22(41-13)28-9-26-14-19(23)24-8-25-20(14)28/h2-5,8-9,12-13,15-18,21-22,30-33H,6-7H2,1H3,(H3-,23,24,25,34,35,36,37)/t12-,13-,15-,16-,17-,18-,21-,22-/m1/s1

InChI key

KPVQNXLUPNWQHM-RBEMOOQDSA-N

Linkage

Analog of NADH

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jixu Li et al.
Antimicrobial agents and chemotherapy, 64(3) (2020-01-08)
Toxoplasma gondii is an obligate intracellular protozoan parasite and a successful parasitic pathogen in diverse organisms and host cell types. Hydroxylamine (HYD) and carboxymethoxylamine (CAR) have been reported as inhibitors of aspartate aminotransferases (AATs) and interfere with the proliferation in
J M Bolscher et al.
The Journal of antimicrobial chemotherapy, 70(5), 1357-1366 (2015-02-11)
The development of drugs to reduce malaria transmission is an important part of malaria eradication plans. We set out to develop and validate a combination of new screening assays for prioritization of transmission-blocking molecules. We developed high-throughput assays for screening
Janette Reader et al.
Malaria journal, 14, 213-213 (2015-05-23)
The discovery of malaria transmission-blocking compounds is seen as key to malaria elimination strategies and gametocyte-screening platforms are critical filters to identify active molecules. However, unlike asexual parasite assays measuring parasite proliferation, greater variability in end-point readout exists between different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service