A1582
Azodicarboxylic acid dipiperidide
~99% (TLC)
Synonym(s):
ADD, ADDP, Azodicarboxylic dipiperidide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H20N4O2
CAS Number:
Molecular Weight:
252.31
Beilstein/REAXYS Number:
261917
MDL number:
UNSPSC Code:
12352000
assay
~99% (TLC)
mp
132-136 °C (lit.)
SMILES string
O=C(\N=N\C(=O)N1CCCCC1)N2CCCCC2
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tsunoda, T, et al.
Tetrahedron Letters, 34, 1639-1639 (1993)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service