Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A140

Sigma-Aldrich

Arecaidine propargyl ester hydrobromide

solid

Synonym(s):

1-Methyl-1,2,5,6-tetrahydro-3-pyridine carboxylic acid propargyl ester hydrobromide, APE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2 · HBr
CAS Number:
Molecular Weight:
260.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

color

white

solubility

H2O: soluble

storage temp.

2-8°C

SMILES string

Br.CN1CCC=C(C1)C(=O)OCC#C

InChI

1S/C10H13NO2.BrH/c1-3-7-13-10(12)9-5-4-6-11(2)8-9;/h1,5H,4,6-8H2,2H3;1H

InChI key

LRPUAFALRBYLTA-UHFFFAOYSA-N

Gene Information

human ... CHRM2(1129)

Biochem/physiol Actions

Potent muscarinic acetylcholine receptor agonist exhibiting slight selectivity for M2 receptors; inactive at nicotinic receptors.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Zhang et al.
ACS nano, 13(8), 8997-9004 (2019-07-16)
Magnetism in monolayer (ML) VSe2 has attracted broad interest in spintronics, while existing reports have not reached consensus. Using element-specific X-ray magnetic circular dichroism, a magnetic transition in ML VSe2 has been demonstrated at the contamination-free interface between Co and
Oscar J Valverde-Barrantes et al.
The New phytologist, 215(4), 1562-1573 (2017-04-26)
Fine-root traits play key roles in ecosystem processes, but the drivers of fine-root trait diversity remain poorly understood. The plant economic spectrum (PES) hypothesis predicts that leaf and root traits evolved in coordination. Mycorrhizal association type, plant growth form and
Liang Lü et al.
Current zoology, 66(4), 435-444 (2020-07-04)
Staphylinoidea (Insecta: Coleoptera) is one of the most species-rich groups in animals, but its huge diversity can hardly be explained by the popular hypothesis (co-radiation with angiosperms) that applies to phytophagous beetles. We estimated the evolutionary mode of staphylinoid beetles
Julien Goebel et al.
BMC genomics, 18(1), 460-460 (2017-06-15)
Gene duplication has led to a most remarkable adaptation involved in vertebrates' host-pathogen arms-race, the major histocompatibility complex (MHC). However, MHC duplication history is as yet poorly understood in non-mammalian vertebrates, including birds. Here, we provide evidence for the evolution
Sébastien Renaut et al.
Microorganisms, 8(1) (2020-01-16)
Little is known about establishment success of the arbuscular mycorrhizal fungal (AMF) inocula and their effects on a soil-indigenous community of AMF. In this study, we assessed the effect of introducing Rhizophagus irregularis DAOM-197198 in soil under field condition on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service