Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F8141

Sigma-Aldrich

Fibronectin-like Engineered Protein Polymer-plus genetically engineered

powder, sterile; autoclaved, BioReagent, suitable for cell culture

Synonym(s):

ProNectin® F Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Product F8141 is not currently sold in your country. Contact Technical Service

recombinant

expressed in E. coli

Quality Level

sterility

sterile; autoclaved

product line

BioReagent

form

powder

mol wt

~110 kDa by SDS-PAGE

packaging

pkg of 1 mg

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 1 mg/mL (stable for 12 mo. at RT)

shipped in

ambient

storage temp.

room temp

Application

Recommended for use as a cell culture substratum
at 2-10 μg/cm2.

Other Notes

Positively-charged, protein polymer which incorporates multiple copies of the RGD cell attachment epitope from human fibronectin between repeated structural peptide units.[1]

Linkage

Similar to F5022 but more highly charged polymer which encourages better adhesion in many cell lines.

Reconstitution

Dilutions can be made to a working concentration with balanced salt solution; stable up to 2 days at RT. Coated labware is stable at least 4 months at RT.

Legal Information

ProNectin is a registered trademark of Sanyo Chemical Industries, Ltd

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Receptor-specific serum-free cell attachment using a highly stable engineered protein polymer.
Esty, A.
American Biotechnology Laboratory, 44-44 (1991)
Sara Riccardi et al.
Frontiers in genome editing, 4, 846669-846669 (2022-05-03)
Neuronal differentiation is a complex process whose dysfunction can lead to brain disorders. The development of new tools to target specific steps in the neuronal differentiation process is of paramount importance for a better understanding of the molecular mechanisms involved
Evan A Thackaberry et al.
Investigative ophthalmology & visual science, 58(10), 4274-4285 (2017-08-30)
Poly(lactic-co-glycolic) acid (PLGA) inserts have been successfully developed for the treatment of posterior eye disease as a means of reducing injection frequency of intravitreally administered therapeutics. PLGA microspheres are also of interest for the delivery of intravitreal drugs, since they
Julia Ghelman et al.
Glia, 69(11), 2699-2716 (2021-07-30)
Oligodendroglial progenitor cells (OPCs) are highly proliferative and migratory cells, which differentiate into complex myelin forming and axon ensheathing mature oligodendrocytes during myelination. Recent studies indicate that the oligodendroglial cell population is heterogeneous on transcriptional and functional level depending on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service