Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76025

Sigma-Aldrich

Poly(maleic anhydride-alt-1-dodecene), 3-(Dimethylamino)-1-propylamine derivative

BioReagent, for molecular biology

Synonym(s):

PMAL®-C10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C21H40O3N2)n
CAS Number:
UNSPSC Code:
12352200

grade

for molecular biology

description

zwitterionic

product line

BioReagent

λ

0.1 % in H2O

UV absorption

λ: 260 nm Amax: ≤0.400
λ: 280 nm Amax: ≤0.150

SMILES string

CCCCCCCCCCC(C)CC(C(C)C(=O)NCCCN(C)C)C(O)=O

Application

Use of "Amphipols" for delivering of membrane proteins to lipid bilayers, as activity support of membrane enzymes.

Analysis Note

enzyme traces DNases, RNases, proteases and phosphatases not detectable

Legal Information

PMAL is a registered trademark of Affymetrix, Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J K Nagy et al.
FEBS letters, 501(2-3), 115-120 (2001-07-27)
Data are presented which suggest that a class of amphiphilic polymers known as 'amphipols' may serve as a vehicle for delivering complex integral membrane proteins into membranes. The integral membrane protein diacylglycerol kinase (DAGK) was maintained in soluble form by
Bonnie M Gorzelle et al.
Journal of the American Chemical Society, 124(39), 11594-11595 (2002-09-26)
Amphipathic polymers ("amphipols") were introduced several years ago (Tribet, C.; Audebert, R.; Popot, J.-L. Proc. Natl. Acad. Sci. U.S.A. 1996, 93, 15047-15050) as an alternative method for solubilizing integral membrane proteins in stable, nativelike conformations. However, direct maintenance of full

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service