Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67610

Sigma-Aldrich

1-Methyl-3-indolyl-β-D-galactopyranoside

≥98.0% (HPLC)

Synonym(s):

Green-β-D-gal, N-Methylindolyl-β-D-galactopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO6
CAS Number:
Molecular Weight:
309.31
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

assay

≥98.0% (HPLC)

form

solid

optical activity

[α]20/D −41.5±2°, c = 1% in DMF/H2O 1:1

storage temp.

2-8°C

SMILES string

Cn1cc(O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)c3ccccc13

InChI

1S/C15H19NO6/c1-16-6-10(8-4-2-3-5-9(8)16)21-15-14(20)13(19)12(18)11(7-17)22-15/h2-6,11-15,17-20H,7H2,1H3/t11-,12+,13+,14-,15-/m1/s1

InChI key

FDECOIVNOJUVCV-GZBLMMOJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chromogenic substrate for β-galactosidase producing a green insoluble product.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabine Wislet-Gendebien et al.
PloS one, 7(10), e46425-e46425 (2012-10-17)
Bone marrow stromal cells are adult multipotent cells that represent an attractive tool in cellular therapy strategies. Several studies have reported that in vitro passaging of mesenchymal stem cells alters the functional and biological properties of those cells, leading to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service