Skip to Content
MilliporeSigma
All Photos(1)

Documents

60494

Sigma-Aldrich

Potassium phosphate tribasic monohydrate

BioUltra, ≥95% (T)

Synonym(s):

tert-Potassium phosphate monohydrate, Tripotassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K3O4P · H2O
CAS Number:
Molecular Weight:
230.28
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

product line

BioUltra

assay

≥95% (T)

impurities

insoluble matter, passes filter test
≤0.6% free alkali

loss

≤13% loss on ignition, 800 °C

pH

12.5-14.0 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤1000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.02

SMILES string

O.[K+].[K+].[K+].[O-]P([O-])([O-])=O

InChI

1S/3K.H3O4P.H2O/c;;;1-5(2,3)4;/h;;;(H3,1,2,3,4);1H2/q3*+1;;/p-3

InChI key

RMNIZOOYFMNEJJ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service