Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59965

Sigma-Aldrich

Nonyl β-D-maltoside

≥99.0% (TLC)

Synonym(s):

n-Nonyl β-D-maltopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H40O11
CAS Number:
Molecular Weight:
468.54
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

assay

≥99.0% (TLC)

mol wt

468.54 g/mol

solubility

H2O: 10 mg/mL, clear, colorless

SMILES string

CCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

InChI

1S/C21H40O11/c1-2-3-4-5-6-7-8-9-29-20-18(28)16(26)19(13(11-23)31-20)32-21-17(27)15(25)14(24)12(10-22)30-21/h12-28H,2-11H2,1H3/t12-,13-,14-,15+,16-,17-,18-,19-,20-,21-/m1/s1

InChI key

KCCBGPCYGBPHBR-ZESVGKPKSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takao Hibi et al.
Biochemical and biophysical research communications, 374(3), 465-469 (2008-07-22)
The inward-rectifying K(+) channel KAT1 is expressed mainly in Arabidopsis thaliana guard cells. The purification of functional KAT1 has never been reported. We investigated the extraction of the plant K(+) channel KAT1 with different detergents, as an example for how
M Gabriella Santonicola et al.
Biophysical journal, 94(9), 3647-3658 (2008-02-01)
The binding of alkyl polyglucoside surfactants to the integral membrane protein bacteriorhodopsin (BR) and the formation of protein-surfactant complexes are investigated by sedimentation equilibrium via analytical ultracentrifugation and by small-angle neutron scattering (SANS). Contrast variation techniques in SANS enable measurement
null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service