Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55845

Sigma-Aldrich

1,2-Distearoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

≥97.0% (TLC)

Synonym(s):

1,2-Dioctadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, 1,2-Distearoyl-sn-glycero-3-phosphoglycerol, L-α-Phosphatidyl-DL-glycerol, distearoyl, DSPG-Na, Octadecanoic acid (1R)-1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester monosodium salt, PG(18:0/18:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NaO10P
CAS Number:
Molecular Weight:
801.06
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

assay

≥97.0% (TLC)

form

powder

functional group

ester

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C42H83O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h39-40,43-44H,3-38H2,1-2H3,(H,47,48);/q;+1/p-1/t39?,40-;/m1./s1

InChI key

YNQYZBDRJZVSJE-QTOMIGAPSA-M

Biochem/physiol Actions

DSPG is the most efficient phosphatidylglycerol in enhancing respiratory mucus clearance in cystic fibrosis.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Cunha et al.
FEBS letters, 469(2-3), 159-162 (2000-03-14)
Kainate receptors are ionotropic receptors, also reported to couple to G(i)/G(o) proteins, increasing neuronal excitability through disinhibition of neuronal circuits. We directly tested in hippocampal synaptosomes if kainate receptor-mediated inhibition of GABA release involved a metabotropic action. The kainate analogue
Reagan L Pennock et al.
Journal of neurophysiology, 115(5), 2376-2388 (2016-02-26)
Whereas the activation of Gαi/o-coupled receptors commonly results in postsynaptic responses that show acute desensitization, the presynaptic inhibition of transmitter release caused by many Gαi/o-coupled receptors is maintained during agonist exposure. However, an exception has been noted where GABAB receptor
Oksana Lenina et al.
Experimental physiology, 104(7), 1004-1010 (2019-05-11)
What is the central question of this study? Do GABA receptors play any role at the neuromuscular junction? What is the main finding and its importance? In the presence of either ionotropic or metabotropic GABA receptor antagonists, diaphragm muscle force
Sarah Y Branch et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(34), 13861-13872 (2013-08-24)
Restriction of food intake increases the acquisition of drug abuse behavior and enhances the reinforcing efficacy of those drugs. However, the neurophysiological mechanisms responsible for the interactions between feeding state and drug use are largely unknown. Here we show that
S Girod de Bentzmann et al.
The European respiratory journal, 6(8), 1156-1161 (1993-09-01)
We have previously shown that a decreased level of phosphatidylglycerol in cystic fibrosis (CF) respiratory mucus is partly responsible for its marked adhesiveness and stickiness, which impair mucus transport, and that distearoyl phosphatidylglycerol (DSPG) was the most efficient form of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service