Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53892

Sigma-Aldrich

4-Hydrazino-7-nitro-benzofurazan hydrazine adduct

suitable for fluorescence

Synonym(s):

7-Nitro-4-benzofurazanyl-hydrazine hydrazine adduct, NBD-H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N5O3 · NH2NH2
CAS Number:
Molecular Weight:
227.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

fluorescence

λex 445 nm; λem 522 nm in chloroform (after derivatization with formaldehyde)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

NN.NNc1ccc([N+]([O-])=O)c2nonc12

InChI

1S/C6H5N5O3.H4N2/c7-8-3-1-2-4(11(12)13)6-5(3)9-14-10-6;1-2/h1-2,8H,7H2;1-2H2

InChI key

BLIOAKMSUDUCKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Hydrazino-7-nitro-benzofurazan (HBD) is used as a fluorescent reagent for protein labeling and pre-column derivatization for chromatographic procedures such as HPLC.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Fluorometric detection of aliphatic aldehydes[1]

pictograms

Flame

signalword

Warning

hcodes

pcodes

Hazard Classifications

Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao-Wen Wu et al.
Chembiochem : a European journal of chemical biology, 12(18), 2813-2821 (2011-11-17)
Rab GTPases play a key role in the regulation of membrane trafficking. Post-translational geranylgeranylation is critical for their biological activity and is conferred by Rab geranylgeranyl transferease (RabGGTase), together with an accessory factor, Rab escort protein (REP). Mechanistic studies of
H. Koizumi et al.
Journal of Chromatography A, 457, 299-299 (1988)
M J Hinojo et al.
Food microbiology, 23(2), 119-127 (2006-09-01)
Fumonisin B1 (FB1) and fumonisin B2 (FB2) are the main members of a family of mycotoxins produced by various fungal species belonging to the Gibberella fujikuroi complex. The present work shows the results of a comparative study of various clean-up
Taeko Mizutani et al.
Journal of dermatological science, 84(3), 314-321 (2016-10-17)
Carbonylated proteins (CPs) are generated by the reaction of basic amino acid residues in proteins with aldehyde compounds produced during lipid peroxidation. CPs in the stratum corneum (SC) impact skin conditions such as skin moisture functions including water content and
H W Lee et al.
Journal of biochemistry, 127(6), 1053-1056 (2000-06-01)
Growth of Escherichia coli in the presence of glyphosate, an inhibitor of aromatic amino acid biosynthesis, has permitted the production of proton-dislocating ATPase that is specifically labeled with 5-fluorotryptophan. Five sets of (19)F resonances could be assigned to each tryptophan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service