Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

51980

Sigma-Aldrich

n-Heptyl β-D-glucopyranoside

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H26O6
CAS Number:
Molecular Weight:
278.34
Beilstein/REAXYS Number:
8806
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

assay

≥98.0% (TLC)

optical activity

[α]20/D −33±2°, c = 5% in H2O

mol wt

278.34 g/mol

technique(s)

RNA extraction: suitable
protein quantification: suitable

CMC

79

SMILES string

CCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C13H26O6/c1-2-3-4-5-6-7-18-13-12(17)11(16)10(15)9(8-14)19-13/h9-17H,2-8H2,1H3/t9-,10-,11+,12-,13-/m1/s1

InChI key

NIDYWHLDTIVRJT-UJPOAAIJSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Non-ionic detergent used for solubilizing membrane proteins[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

n-Alkyl-glucosides as detergents for the preparation of highly homogeneous bilayer liposomes of variable sizes ( 60-240 nm phi) applying defined rates of detergent removal by dialysis.
R A Schwendener et al.
Biochemical and biophysical research communications, 100(3), 1055-1062 (1981-06-16)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service