Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44559

Sigma-Aldrich

2-O-(2-Acetamido-2-deoxy-β-D-glucopyranosyl)-D-mannose

≥98.0% (TLC)

Synonym(s):

N-Acetylglucosaminyl-β-1,2-mannose, β-D-GlcNAc-(1→2)-D-Man

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25NO11
CAS Number:
Molecular Weight:
383.35
UNSPSC Code:
12352201
NACRES:
NA.25

assay

≥98.0% (TLC)

suitability

conforms to structure for Infrared spectrum

storage temp.

−20°C

SMILES string

CC(N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1O[C@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO)=O

InChI

1S/C14H25NO11/c1-5(19)15-9-13(24)12(23)8(4-18)26-14(9)25-7(3-17)11(22)10(21)6(20)2-16/h3,6-14,16,18,20-24H,2,4H2,1H3,(H,15,19)/t6-,7-,8-,9-,10-,11-,12-,13-,14-/m1/s1

InChI key

WCIQTQUMKQRLKV-ZTGOBPNGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Disaccharide fragment found in N-acetyl-lactosaminic type glycans of N-glycoproteins.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tammy T Fang et al.
Journal of the American Chemical Society, 129(31), 9721-9736 (2007-07-17)
Mass spectrometry of hexose-containing disaccharides often yields product ions of m/z 221 in the negative ion mode. Using a Paul trap, isolation and collision-induced dissociation of the m/z 221 anions yielded mass spectra that easily differentiated their stereochemistry and anomeric
Velavan Ramasamy et al.
Journal of molecular biology, 353(1), 53-67 (2005-09-15)
beta-1,4-Galactosyltransferase-I (beta4Gal-T1) transfers galactose from UDP-galactose to N-acetylglucosamine (GlcNAc) residues of the branched N-linked oligosaccharide chains of glycoproteins. In an N-linked biantennary oligosaccharide chain, one antenna is attached to the 3-hydroxyl-(1,3-arm), and the other to the 6-hydroxyl-(1,6-arm) group of mannose
Benjamin Pluvinage et al.
Structure (London, England : 1993), 19(11), 1603-1614 (2011-11-15)
The complete degradation of N-linked glycans by the pathogenic bacterium Streptococcus pneumoniae is facilitated by the large multimodular cell wall-attached exo-β-D-N-acetylglucosaminidase StrH. Structural dissection of this virulence factor using X-ray crystallography showed it to have two structurally related glycoside hydrolase
Yong-Liang Jiang et al.
The Journal of biological chemistry, 286(50), 43004-43012 (2011-10-21)
The β-N-acetylhexosaminidase (EC 3.2.1.52) from glycoside hydrolase family 20 (GH20) catalyzes the hydrolysis of the β-N-acetylglucosamine (NAG) group from the nonreducing end of various glycoconjugates. The putative surface-exposed N-acetylhexosaminidase StrH/Spr0057 from Streptococcus pneumoniae R6 was proved to contribute to the
Zoi Pipirou et al.
Glycobiology, 21(6), 806-812 (2011-01-25)
The biochemical properties of mouse LSECtin, a glycan-binding receptor that is a member of the C-type lectin family found on sinusoidal endothelial cells, have been investigated. The C-type carbohydrate-recognition domain of mouse LSECtin, expressed in bacteria, has been used in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service