Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39133

Sigma-Aldrich

D-Erythrono-1,4-lactone

≥95.0% (GC)

Synonym(s):

D-Erythronic acid γ-lactone, (3R-cis) (−)-Dihydro-3,4-dihydroxy-2(3H)-furanone, D-Erythronolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O4
CAS Number:
Molecular Weight:
118.09
Beilstein/REAXYS Number:
81073
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

assay

≥95.0% (GC)

optical activity

[α]/D -71.0±3.0°, c = 1 in H2O

mp

100-102 °C (lit.)

SMILES string

O[C@@H]1COC(=O)[C@@H]1O

InChI

1S/C4H6O4/c5-2-1-8-4(7)3(2)6/h2-3,5-6H,1H2/t2-,3-/m1/s1

InChI key

SGMJBNSHAZVGMC-PWNYCUMCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon-branched carbohydrate chirons: practical access to both enantiomers of 2-C-methyl-ribono-1,4-lactone and 2-C-methyl-arabinonolactone.
Booth, K. V., et al.
Tetrahedron Asymmetry, 20, 2357-2367 (2009)
Synthesis of ?-Substituted 3-Ulosonic Acids from Aldonolactones.
Csuk, R., et al.
Tetrahedron, 59, 7887-7895 (2003)
Viktoriia Korniienko et al.
Materials (Basel, Switzerland), 13(18) (2020-09-10)
High strength, excellent corrosion resistance, high biocompatibility, osseointegration ability, and low bacteria adhesion are critical properties of metal implants. Additionally, the implant surface plays a critical role as the cell and bacteria host, and the development of a simultaneously antibacterial
H S Bleeg et al.
European journal of biochemistry, 127(2), 391-396 (1982-10-01)
An enzyme from Saccharomyces cerevisiae which catalyzes the reaction: L-galactonolactone + O2 leads to L-ascorbate + H2O2 has been purified 466-fold from the mitochondrial fraction of a yeast homogenate. The enzyme has several properties that are different from the L-galactonolactone
Biosynthesis of Vitamin B6: Origin of Pyridoxine by the Union of Two Acyclic Precursors, 1-Deoxy-D-xylulose and 4-Hydroxy-L-threonine.
Kennedy, I.A., et al.
Journal of the American Chemical Society, 117, 1661-1662 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service