Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38216

Sigma-Aldrich

N-[4-(5,6-Dimethoxy-N-phthalimidinyl)phenyl]maleimide

≥97.0% (HPLC)

Synonym(s):

DPM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16N2O5
CAS Number:
Molecular Weight:
364.35
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥97.0% (HPLC)

SMILES string

COc1cc2CN(C(=O)c2cc1OC)c3ccc(cc3)N4C(=O)C=CC4=O

InChI

1S/C20H16N2O5/c1-26-16-9-12-11-21(20(25)15(12)10-17(16)27-2)13-3-5-14(6-4-13)22-18(23)7-8-19(22)24/h3-10H,11H2,1-2H3

Inchi Key

ISCHQGBNSNVWCQ-UHFFFAOYSA-N

Application

N-[4-(5,6-Dimethoxy-N-phthalimidinyl)phenyl]maleimide (DPM) is used to fluorescence label proteins via their sulfhydryl groups. Nonfluorescent N-substituted maleimides react selectively with thiol compounds to form fluorescent derivatives. Fluorescent intensity of the dimethoxyphthalimidyl group is higher than that of the non substituted phthalimidyl group.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mike Goblirsch et al.
PloS one, 8(3), e58165-e58165 (2013-03-14)
Persistent exposure to mite pests, poor nutrition, pesticides, and pathogens threaten honey bee survival. In healthy colonies, the interaction of the yolk precursor protein, vitellogenin (Vg), and endocrine factor, juvenile hormone (JH), functions as a pacemaker driving the sequence of
Aude Picard et al.
Frontiers in microbiology, 2, 263-263 (2011-12-31)
Microbial heterotrophic activity was investigated in oxic sub-seafloor sediments at North Pond, a sediment pond situated at 23°N on the western flank of the Mid-Atlantic Ridge. The North Pond sediments underlie the oligotrophic North Atlantic Gyre at 4580-m water depth
P N McCormick et al.
Neuropharmacology, 89, 225-231 (2014-10-07)
Pramipexole is a clinically important dopamine receptor agonist with reported selectivity for dopamine D3 receptors over other dopaminergic and non-dopaminergic sites. Many of its behavioural effects are therefore attributed to D3 receptor activity. Here we relate pramipexole's ex vivo D2 and
Vanessa Malagnino et al.
Molecular pharmacology, 96(2), 128-137 (2019-05-28)
Drug transporters play a crucial role in pharmacokinetics. One subfamily of transporters with proven clinical relevance are the OATP1B transporters. Recently we identified a new member of the OATP1B family named OATP1B3-1B7 (LST-3TM12). This functional transporter is encoded by SLCO1B3
Eric J Esswein et al.
Journal of occupational and environmental hygiene, 15(1), 63-70 (2017-10-21)
Diesel engines serve many purposes in modern oil and gas extraction activities. Diesel particulate matter (DPM) emitted from diesel engines is a complex aerosol that may cause adverse health effects depending on exposure dose and duration. This study reports on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service